Please login to your online account to display your discounted pricing

Starch Indicator, Preserved with Chloroform (Mercury Free), Certified, 1%, LabChem™

$46.26 - $123.80

Chemical Identifiers

Chemical Name or Material Starch Indicator
CAS 7732-18-5
CAS Max % 98.5
CAS 9005-25-8
CAS Max % 1
CAS 67-66-3
CAS Max % 0.495
CAS 64-17-5
CAS Max % 0.005
Name Note Preserved with Chloroform (Mercury Free)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC253151

 
lab chem
LC253151
Poly Bottle 500mL Each for $46.26

LC253152

 
lab chem
LC253152
Poly Bottle 1L Each for $80.91

LC253154

 
lab chem
LC253154
Poly Bottle 4L Each for $123.80
Description & Specifications

Specifications

Purity Grade Certified Grade
Chemical Name or Material Starch Indicator
Quantity 500mL
CAS Max % 0.495
CAS 64-17-5
CAS Max % 0.005
Name Note Preserved with Chloroform (Mercury Free)
Physical Form Liquid
Assay 1%
CAS 7732-18-5
Identification Passes Test
Health Hazard 1 Warning
Packaging Poly Bottle
CAS Max % 98.5
Health Hazard 2 GHS H Statement
Suspected of causing cancer (Inhalation).
CAS 9005-25-8
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/eye protection/face protection.
If exposed or concerned: Get medical attention.
Store locked up.
Dispose of contents/container in accordance with local, state and federal regulations.
Solubility Soluble in water
Specific Gravity 1g/mL
Suitability Passes Test
CAS Max % 1
Recommended Storage Room Temperature
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Density 1g/mL
CAS 67-66-3
Color Clear-slightly opaque colorless
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/eye protection/face protection.
If exposed or concerned: Get medical attention.
Store locked up.
Dispose of contents/container in accordance with local, state and federal regulations.

GHS H Statement
Suspected of causing cancer (Inhalation).

Room Temperature