Please login to your online account to display your discounted pricing

Starch Indicator, 0.5% (w/v) Aqueous Solution, with Chloroform preservative, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Starch Indicator
Name Note With Chloroform Preservative
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 9005-84-9
CAS Min % 0.49
CAS Max % 0.51
CAS 67-66-3
CAS Min % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

80021

 
ricca chemical company
RI80021
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

800232

 
ricca chemical company
RI800232
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

800216

 
ricca chemical company
RI800216
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

80025

 
ricca chemical company
RI80025
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Translucent
Quantity 4L
CAS 7732-18-5
CAS Max % 100.0
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 0.5% (w/v)
Packaging Poly natural
CAS Min % 98.0
CAS Min % 0.49
CAS 67-66-3
CAS Max % 0.4
CAS Min % 0.1
CAS Max % 0.51
CAS 9005-84-9
Name Note With Chloroform Preservative
Chemical Name or Material Starch Indicator