Please login to your online account to display your discounted pricing

Starch, ACROS Organics™


Quantity 500mL
Packaging Plastic bottle

Manufacturer: fisher scientific acros  AC380645000

 View more versions of this product

Catalog No. AC380645000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Solution
Quantity 500mL
Color Clear to slightly turbid
Density 1g/mL
Chemical Name or Material Starch
Molecular Formula (C6H10O5)n
Packaging Plastic bottle
CAS 7732-18-5
MDL Number MFCD00082026
CAS 9005-25-8
Sensitivity Passes Test

Specifications

Chemical Name or Material Starch
CAS 9005-25-8
CAS 7732-18-5
Purity Grade Extra Pure
Molecular Formula (C6H10O5)n
MDL Number MFCD00082026