Please login to your online account to display your discounted pricing

Stannous Chloride Reagent II, for Phosphate Analysis (with extraction), Ricca Chemical


For Analysis of pHosphate Analysis (With Extraction)

Quantity 1L
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR79980001A

 View more versions of this product

Catalog No. R79980001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Stannous Chloride Reagent II
CAS 56-81-5
CAS Min % 98.0
CAS Max % 100.0
CAS 10025-69-1
CAS Min % 0.26
CAS Max % 0.27
Physical Form Liquid
Packaging Plastic Bottle
Color Clear colorless
Quantity 1L

Specifications

Chemical Name or Material Stannous Chloride Reagent II
CAS 56-81-5
CAS Min % 98.0
CAS Max % 100.0
CAS 10025-69-1
CAS Min % 0.26
CAS Max % 0.27
Physical Form Liquid