Please login to your online account to display your discounted pricing

Stannous Chloride Reagent II, For Phosphorus, LabChem™

$129.66 - $129.66

Chemical Identifiers

Chemical Name or Material Stannous Chloride Reagent II
CAS 56-81-5
CAS Max % 99.6
CAS 10025-69-1
CAS Max % 0.4
Name Note For Phosphorus
Linear Formula SnCl2·2H2O
Formula Weight 225.65
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC252401

 
lab chem
LC252401
500mL Each for $129.66
Description & Specifications

Specifications

Formula Weight 225.65
Chemical Name or Material Stannous Chloride Reagent II
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
Name Note For Phosphorus
Physical Form Liquid
Linear Formula SnCl2·2H2O
CAS 56-81-5
CAS Max % 99.6
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 10025-69-1
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 0.4
Solubility Soluble in water
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.