Please login to your online account to display your discounted pricing

Stannous Chloride Reagent I, for Phosphate Analysis (without extraction), Ricca Chemical

For Analysis of pHosphate (Without Extraction)

$65.06 - $186.12

Chemical Identifiers

Chemical Name or Material Stannous Chloride Reagent I
CAS 56-81-5
CAS Min % 96.0
CAS Max % 100.0
CAS 10025-69-1
CAS Min % 1.9
CAS Max % 1.98
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7997-4

 
ricca chemical company
RI79974
Poly natural 120mL Each for $65.06

7997-16

 
ricca chemical company
RI799716
Poly natural 500mL Each for $186.12

R79970001A

 
ricca chemical company
RIR79970001A
Plastic Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 120mL
CAS Min % 96.0
CAS Max % 100.0
CAS Min % 1.9
CAS Max % 1.98
CAS 56-81-5
Packaging Poly natural
CAS 10025-69-1
Chemical Name or Material Stannous Chloride Reagent I