Please login to your online account to display your discounted pricing

Stannous Chloride Reagent I, For Phosphorus, Certified, LabChem™

$141.66 - $141.66

Chemical Identifiers

Chemical Name or Material Stannous Chloride Reagent I
CAS 56-81-5
CAS Max % 97.5
CAS 10025-69-1
CAS Max % 2.5
Name Note For Phosphorus
Purity Grade Certified Grade
Linear Formula SnCl2·2H2O
Formula Weight 225.65
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC252301

 
lab chem
LC252301
500mL Each for $141.66
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 225.65
Chemical Name or Material Stannous Chloride Reagent I
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
Name Note For Phosphorus
Physical Form Liquid
Linear Formula SnCl2·2H2O
CAS 56-81-5
Health Hazard 1 Warning
CAS Max % 97.5
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
Identification Passes Test
CAS 10025-69-1
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Poly Bottle
CAS Max % 2.5
Solubility Soluble in water
Suitability Passes Test
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation and serious eye irritation.