missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Stannous Chloride, For Arsenic, 40%, LabChem™

Supplier:  LabChem LC252001

 View more versions of this product

Catalog No. LC252001



Description

Description

Specifications

Specifications

Stannous Chloride
For Arsenic
60,40
SnCl2·2H2O
UN1789
Soluble in water
Cl
36.46
CHEBI:17883
40%
Amber Glass
500 mL
Colorless
7647-01-0,10025-69-1
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
225.65
Passes Test
Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Liquid

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection, protective clothing, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center or doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center or doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center or doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center or doctor.
Store locked up in a well-

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions