Please login to your online account to display your discounted pricing

Stannous Chloride Dihydrate (Crystalline/Technical), Fisher Chemical

$474.07 - $474.07

Chemical Identifiers

Chemical Name or Material Stannous Chloride Dihydrate
CAS 10025-69-1
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Cl2Sn · 2 H2O
Formula Weight 225.65
MDL Number MFCD00149863
Synonym Tin (II) Chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

T163-500

  SDS
fisher scientific company
T163500
Poly Bottle 500g Each for $474.07
Case of 6 Each for $2,844.42
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 225.65
Boiling Point 652°C
Melting Point 37°C
Quantity 500g
DOT Information DOT Class 8, : Corrosive
Molecular Formula Cl2Sn · 2 H2O
Color Colorless
Physical Form Solid
Viscosity [NULL]
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. May cause an allergic skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1
Identification Pass Test
MDL Number MFCD00149863
CAS 10025-69-1
ChemAlert Storage Symbol White
Synonym Tin (II) Chloride
Chemical Name or Material Stannous Chloride Dihydrate
Packaging Poly Bottle
Health Hazard 2 DANGER!

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Causes burns by all exposure routes. May cause an allergic skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1

DANGER!