Please login to your online account to display your discounted pricing

Stannous Chloride, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$637.73 - $637.73

Chemical Identifiers

Chemical Name or Material Stannous Chloride
CAS 10025-69-1
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula SnCl2·2H2O
Formula Weight 225.63
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004317

 
avantor performance
398001
Poly Bottle 500g Each for $637.73

02004318

 
avantor performance
398005
Poly Bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 225.63
Purity Grade Notes Meets ACS reagent chemical requirements
Chemical Name or Material Stannous Chloride
Molecular Formula SnCl2·2H2O
Name Note Dihydrate
Packaging Poly Bottle
Specific Gravity 1L = 2.71kg
CAS 10025-69-1
Quantity 500g

    Suitable for Mercury Determination