Please login to your online account to display your discounted pricing

Stannous Chloride, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  398005

 View more versions of this product

Catalog No. 02004318

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 225.63
Purity Grade Notes Meets ACS reagent chemical requirements
Chemical Name or Material Stannous Chloride
Name Note Dihydrate
Packaging Poly Bottle
Specific Gravity 1L = 2.71kg
Molecular Formula SnCl2·2H2O
CAS 10025-69-1
Quantity 2.5kg

Specifications

Chemical Name or Material Stannous Chloride
CAS 10025-69-1
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula SnCl2·2H2O
Formula Weight 225.63

    Suitable for Mercury Determination