Please login to your online account to display your discounted pricing

Stannous Chloride, Dihydrate, ACS Grade, 98.0% to 103.0%, LabChem™

$204.11 - $204.11

Chemical Identifiers

Chemical Name or Material Stannous Chloride
CAS 10025-69-1
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula SnCl2·2H2O
Formula Weight 225.65
UN Number UN3260
Synonym Tin Chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC251701

 
lab chem
LC251701
500g Each for $204.11
Description & Specifications

Specifications

Assay 98.0 to 103.0%
Formula Weight 225.65
Chemical Name or Material Stannous Chloride
Identification Passes Test
Name Note Dihydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Specific Gravity 2710kg/m3
Storage Information Room Temperature
Linear Formula SnCl2·2H2O
CAS 10025-69-1
UN Number UN3260
Decomposition Information Hydrogen chloride
Synonym Tin Chloride
Color White
Melting Point 38°C
Quantity 500g
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Do not breathe dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2,710kg/m3
Safety and Handling

Danger

GHS P Statement
Do not breathe dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.