Please login to your online account to display your discounted pricing

Stannous Chloride, Certified, 10% (w/v), LabChem™

$180.00 - $180.00

Chemical Identifiers

Chemical Name or Material Stannous Chloride
CAS 7732-18-5
CAS Max % 81.5
CAS 10025-69-1
CAS Max % 10
CAS 7647-01-0
CAS Max % 8.5
Purity Grade Certified Grade
Linear Formula SnCl2·2H2O
Formula Weight 225.65
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC251801

 
lab chem
LC251801
500mL Each for $180.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 225.65
Chemical Name or Material Stannous Chloride
Synonym Tin Chloride
Health Hazard 1 Danger
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 10
Quantity 500mL
Identification Passes Test
CAS 10025-69-1
Physical Form Liquid
CAS 7647-01-0
UN Number UN1789
CAS Max % 8.5
Suitability Passes Test
CAS Max % 81.5
CAS 7732-18-5
Color Clear, colorless
Packaging Amber Glass
Linear Formula SnCl2·2H2O
Solubility Soluble in water
Storage Information Room Temperature
Assay 10% (w/v)
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.