Please login to your online account to display your discounted pricing

Stannous Chloride AR Crystal, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging STAKMOR

Manufacturer: avantor performance  817620

 View more versions of this product

Catalog No. MK8176A12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging STAKMOR
Quantity 12kg
Chemical Name or Material Stannous Chloride
Name Note Dihydrate
CAS 10025-69-1
Physical Form Crystalline
Molecular Formula SnCl2·2H2O
Formula Weight 225.63g/mol

Specifications

Chemical Name or Material Stannous Chloride
Name Note Dihydrate
CAS 10025-69-1
Physical Form Crystalline
Molecular Formula SnCl2·2H2O
Formula Weight 225.63g/mol