Please login to your online account to display your discounted pricing

Stannous Chloride Anhydrous (Crystalline Powder or Flakes/Technical), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  T141500

 View more versions of this product

Catalog No. T141-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Boiling Point 652°C
Formula Weight 189.62
Melting Point 246°C
Quantity 500g
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
pH 2
Color White
Density 3.950g/cm³
Molecular Formula Cl2Sn
Chemical Name or Material Stannous Chloride Anhydrous
CAS 7772-99-8
MDL Number MFCD00011241
Synonym Tin (II) Chloride Anhydrous
Identification Pass Test
Packaging Glass Bottle
ChemAlert Storage Symbol White

Specifications

Chemical Name or Material Stannous Chloride Anhydrous
CAS 7772-99-8
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Cl2Sn
Formula Weight 189.62
MDL Number MFCD00011241
Synonym Tin (II) Chloride Anhydrous
Accessories
 

SDS & Safety Documents