Standard Reference Solution, pH 7.0 Buffer, Macron Fine Chemicals™
macron
Manufacturer:  Avantor Performance 003110
 View more versions of this product
Catalog No. MK00314
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Clear, colorless solution
6.96 to 7.04
4L
Liquid
Yes
Chemical Identifiers
0.0
icon-pdf.png
Documents