Please login to your online account to display your discounted pricing

Standard Reference Solution, pH 7.0 Buffer, Macron Fine Chemicals™

macron

Chemical Identifiers

Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MK0031500

 
avantor performance
003104
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK00314

 
avantor performance
003110
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Physical Form Liquid
Appearance Clear, colorless solution
pH Range 6.96 to 7.04
Traceability to NIST Yes