Please login to your online account to display your discounted pricing

Standard Reference Solution, pH 7.0 Buffer, Color Coded Yellow, Macron Fine Chemicals™

macron

Chemical Identifiers

Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

14650354

 
avantor performance
009810
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Appearance Clear, yellow solution
Physical Form Liquid
pH Range 6.96 to 7.04
Traceability to NIST Yes
Quantity 4L