Standard Reference Solution, pH 4.01 Buffer, Color Coded Red, Macron Fine Chemicals™
macron
Manufacturer:  Avantor Performance 009704
 View more versions of this product
Catalog No. MK0097500
Specifications
Specifications
Clear, red solution
4.00 to 4.02
500mL
Liquid
Yes
Chemical Identifiers
0.0
icon-pdf.png
Documents