Please login to your online account to display your discounted pricing

Standard Reference Solution, pH 4.01 Buffer, Color Coded Red, Macron Fine Chemicals™

macron

Chemical Identifiers

Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

14650402

 
avantor performance
009710
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK0097500

 
avantor performance
009704
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Physical Form Liquid
Appearance Clear, red solution
pH Range 4.00 to 4.02
Traceability to NIST Yes