Please login to your online account to display your discounted pricing

Standard mOsm Solutions, Precision Systems


Quantity 125mL

Manufacturer: precision systems inc  PSY2109

 View more versions of this product

Catalog No. 12-827-16

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 125mL
Physical Form Liquid
Chemical Name or Material Osmometry Standard

Specifications

Chemical Name or Material Osmometry Standard
Physical Form Liquid