Please login to your online account to display your discounted pricing
Stanbio Ser-T-Fy™ I/II Control Assayed Serum
A control serum to monitor the accuracy and precision of manual and automated, clinical chemistry methods
$297.49 - $297.49
Specifications
For Use With (Application) In-vitro diagnostic use only
Includes Therapeutic drugs
Quantity 6 x 5mL
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
22-022-600
stanbio laboratory inc
G42786
Ser-T-Fy™ I normal control Serum Pack of 6 Each for $297.49
22-022-601
stanbio laboratory inc
G42886
Ser-T-Fy™ II abnormal control Serum Pack of 6 Each for $297.49
Description & Specifications
Specifications
For Use With (Application) In-vitro diagnostic use only
Includes Therapeutic drugs
Quantity 6 x 5mL
  • Offers a stable, lyophilized product prepared from human serum with added chemicals
  • Over 60 different constituents including therapeutic drugs
  • For each constituent, assigned values were obtained with Stanbio test reagents; differences in chemistry methodologies and manufacture may result in different expected ranges
  • Reported with Instrument Specific Values