Please login to your online account to display your discounted pricing

Stanbio Laboratory STAT-Site™ Mβ-HB Meter


Measure β-hydroxybutyrate quickly and quantitatively

Manufacturer: stanbio laboratory inc  400400

 View more versions of this product

Catalog No. SB400400

  • / Each
 

Description & Specifications

Specifications

Product Type Ketone meter
For Use With (Application) Determination of beta-Hydroxybutyrate in Serum or Plasma, aiding in the diagnosis and monitoring of ketoacidosis
Calibration Automatic: code key
Sample Size 10μL
Test Time <80 sec.
Precision Low control: 6.1% CV/High control: 3.9% CV
Quantity Each
Sample Type Serum or Plasma
Linearity 0.1 to 2mm