Please login to your online account to display your discounted pricing

Stainless Steel Wire, 1.0mm (0.04 in.) dia., Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Stainless Steel
Physical Form Wire, 1.0mm (0.04 in.) dia.
Molecular Formula Fe:Cr:Ni; 70:19:11 wt%
MDL Number MFCD00148499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40947BW

 
alfa aesar
40947BW
50cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40947BZ

 
alfa aesar
40947BZ
150cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50cm
Color Silver
Chemical Name or Material Stainless Steel
Odor Odorless
Physical Form Wire, 1.0mm (0.04 in.) dia.
Molecular Formula Fe:Cr:Ni; 70:19:11 wt%
MDL Number MFCD00148499
Appearance Silver -colored wire