Upchurch Scientific™ High-Pressure Unions
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
Supplier: Upchurch Scientific™ UPU322
Specifications
Union | |
0.103 μL | |
0.020 in. Thru Hole |
0.02 in. | |
Stainless Steel |