Please login to your online account to display your discounted pricing

Alfa Aesar™ Stainless Steel tubing, 1.65mm (0.065in) OD, 1.19mm (0.047in) ID, Type 304

Chemical Identifiers

Chemical Name or Material Stainless Steel tubing, Type 304
Name Note OD: 1.65mm (0.065 in.), ID: 1.19mm (0.047 in.)
MDL Number MFCD00148499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13469BV

  SDS
alfa aesar™
13469BV
30cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13469CE

  SDS
alfa aesar™
13469CE
5 x 30cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 30cm
Chemical Name or Material Stainless Steel tubing, Type 304
TSCA TSCA
Name Note OD: 1.65mm (0.065 in.), ID: 1.19mm (0.047 in.)
MDL Number MFCD00148499
Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni; 70:19:11 wt%