missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Stainless Steel tubing, 0.82mm (0.032in) OD, 0.51mm (0.02in) ID, Type 304

Chemical Identifiers

MDL Number MFCD00148499
Chemical Name or Material Stainless Steel tubing, Type 304, OD: 0.82mm (0.032 in.), ID: 0.51mm (0.02 in.)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13468BV SDS Alfa Aesar™
13468BV
30cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA13468CE SDS Alfa Aesar™
13468CE
5 x 30cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00148499
Stainless Steel tubing, Type 304, OD: 0.82mm (0.032 in.), ID: 0.51mm (0.02 in.)

Specifications

Fe:Cr:Ni; 70:19:11 wt%
30cm
MFCD00148499
Stainless Steel tubing, Type 304, OD: 0.82mm (0.032 in.), ID: 0.51mm (0.02 in.)

Safety and Handling

TSCA : TSCA