missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Stainless Steel rod, 3.175mm (0.125in) dia, Type 303, Thermo Scientific Chemicals
$100.66 - $100.66
Specifications
Chemical Name or Material | Stainless Steel rod, Type 303 |
---|---|
Name Note | 3.175mm (0.125 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni; 73:18:9 wt% |
MDL Number | MFCD00148499 |
Form | Rod |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA13475RM
|
Thermo Scientific Chemicals
013475RM |
90 cm |
Each for $100.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel rod, Type 303 | |
Fe:Cr:Ni; 73:18:9 wt% | |
Rod |
3.175mm (0.125 in.) dia. | |
MFCD00148499 |
RUO – Research Use Only