Please login to your online account to display your discounted pricing

Alfa Aesar™ Stainless Steel powder, -325 mesh, Type 430-L

Chemical Identifiers

Chemical Name or Material Stainless Steel
Name Note Type 430-L
MDL Number MFCD00148499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4729036

 
alfa aesar™
4729036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4729022

 
alfa aesar™
4729022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47290A3

 
alfa aesar™
47290A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Stainless Steel
TSCA TSCA
Name Note Type 430-L
Physical Form Powder
MDL Number MFCD00148499
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H319-H335
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Si:C; 82.9:16.5:0.5:0.02 wt%
Mesh Size −325 Mesh
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H319-H335
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.