Alfa Aesar™ Stainless Steel powder, -325 mesh, Type 430-L
Manufacturer:  Alfa Aesar™ 4729036
 View more versions of this product
Catalog No. AA4729036
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Fe:Cr:Si:C; 82.9:16.5:0.5:0.02 wt%
−325 Mesh
Type 430-L
Powder
Stainless Steel
500g
MFCD00148499
Documents