Please login to your online account to display your discounted pricing

Alfa Aesar™ Stainless Steel powder, -100 mesh, annealed, Type 410-L

Chemical Identifiers

Chemical Name or Material Stainless Steel powder, Type 410-L
Name Note Annealed
MDL Number MFCD00148499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1108836

  SDS
alfa aesar™
1108836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11088A3

  SDS
alfa aesar™
11088A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Stainless Steel powder, Type 410-L
TSCA TSCA
Name Note Annealed
Physical Form Powder
MDL Number MFCD00148499
Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr; 87.5:12.5 wt%
Mesh Size −100 Mesh