Please login to your online account to display your discounted pricing

Stainless-Steel gauze, Type 304, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Stainless Steel gauze
Name Note Open area: 36%; Width of opening: 0.003 in.
Physical Form Wire Cloth
Molecular Formula Fe:Cr:Ni; 70:19:11 wt.%
MDL Number MFCD00148499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13476CH

 
alfa aesar
13476CH
20 x 30cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13476RR

 
alfa aesar
13476RR
10 x 10cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Wire Cloth
Quantity 20 x 30cm
Chemical Name or Material Stainless Steel gauze
Name Note Open area: 36%; Width of opening: 0.003 in.
Molecular Formula Fe:Cr:Ni; 70:19:11 wt.%
MDL Number MFCD00148499
Health Hazard 1 H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
Safety and Handling

H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.