Alfa Aesar™ Stainless Steel gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire, Type 304
Manufacturer:  Alfa Aesar™ 13478CH
 View more versions of this product
Catalog No. AA13478CH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Fe:Cr:Ni; 70:19:11 wt%
40 mesh
Woven from 0.25mm (0.01 in.) dia. wire
Wire Cloth
Stainless Steel gauze, Type 304
20 x 30cm
MFCD00148499
Documents