Please login to your online account to display your discounted pricing

Alfa Aesar™ Stainless Steel flake, -325 mesh, 0.4-1.2 micron thick, Type 316-L

Chemical Identifiers

Chemical Name or Material Stainless Steel flake, Type 316-L
MDL Number MFCD00148499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1378730

  SDS
alfa aesar™
1378730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13787A1

  SDS
alfa aesar™
13787A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 250g
Chemical Name or Material Stainless Steel flake, Type 316-L
TSCA TSCA
Physical Form Flakes
MDL Number MFCD00148499
Particle Size 0.4 to 1.2 Micron thick
Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Mesh Size −325 Mesh