Please login to your online account to display your discounted pricing

ST 91, Tocris

α2 agonist, putative α2C agonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material ST 91
Assay Percent Range >99%
Molecular Formula C13H19N3·HCl
Formula Weight 253.77
Synonym N-(2, 6-Diethylphenyl)-4, 5-dihydro-1H-imidazol-2-amine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

263810

 
r & d systems
RDS263810
10mg Each for $177.37

263850

 
r & d systems
RDS263850
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 253.77
Quantity 10mg
Chemical Name or Material ST 91
Recommended Storage Desiccate at +4°C
Molecular Formula C13H19N3·HCl
Assay Percent Range >99%
CAS 4749-61-5
Synonym N-(2, 6-Diethylphenyl)-4, 5-dihydro-1H-imidazol-2-amine hydrochloride
Safety and Handling

Desiccate at +4°C