Please login to your online account to display your discounted pricing

ST 1936 oxalate, Tocris™

Selective, high affinity 5-HT6 agonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material ST 1936 oxalate
Assay Percent Range >98%
Molecular Formula C13H17ClN2·C2H2O4
Formula Weight 326.78
Synonym 5-Chloro-3-[2-(dimethylamino)ethyl]-2-methylindole oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

433750

 
r & d systems
433750
50mg Each for $924.23

433710

 
r & d systems
433710
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 326.78
Quantity 50mg
Chemical Name or Material ST 1936 oxalate
Assay Percent Range >98%
Molecular Formula C13H17ClN2·C2H2O4
CAS 1210-81-7
Synonym 5-Chloro-3-[2-(dimethylamino)ethyl]-2-methylindole oxalate
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C