missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ SSPE (20X)

Specifications

Chemical Name or Material SSPE
MDL Number MFCD00132372
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61214K2 View Documents Alfa Aesar™
J61214K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61214K3 View Documents Alfa Aesar™
J61214K3
2L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61214K7 View Documents Alfa Aesar™
J61214K7
4L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

SSPE
MFCD00132372