Please login to your online account to display your discounted pricing

SSEA-4 Mouse anti-Human, eFluor 570, Clone: eBioMC-813-70 (MC-813-70), eBioscience™

Mouse Monoclonal Antibody

$160.95 - $397.30

Specifications

Antigen SSEA-4
Clone eBioMC-813-70 (MC-813-70)
Host Species Mouse
Gene Alias stage-specific embryonic antigen-4
Species Reactivity Canine, Chicken, Human, Mouse, Non-human primate, Rabbit
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123638

 
life technologies
LS41884380
25μg Each for $160.95

501123639

 
life technologies
LS41884382
100μg Each for $397.30
Description & Specifications

Specifications

Antigen SSEA-4
Clone eBioMC-813-70 (MC-813-70)
Host Species Mouse
Gene Alias stage-specific embryonic antigen-4
Species Reactivity Canine, Chicken, Human, Mouse, Non-human primate, Rabbit
Applications Immunocytochemistry
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells.

The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys.