Please login to your online account to display your discounted pricing

SSEA-3 Rat anti-Human, Mouse, PE, Clone: eBioMC-631 (MC-631), eBioscience™

Rat Monoclonal Antibody

$187.05 - $419.05

Specifications

Antigen SSEA-3
Clone eBioMC-631 (MC-631)
Host Species Rat
Gene Alias stage-specific embryonic antigen-3
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011219

 
life technologies
LS12883341
25 tests Each for $187.05

5011220

 
life technologies
LS12883342
100 tests Each for $419.05
Description & Specifications

Specifications

Antigen SSEA-3
Clone eBioMC-631 (MC-631)
Host Species Rat
Gene Alias stage-specific embryonic antigen-3
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The eBioMC-631 (MC-631) monoclonal antibody reacts with the carbohydrate moiety galactosylgloboside on Stage-specific embryonic antigen-3 (SSEA-3). Expression is restricted to the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES) and down regulated following differentiation.