missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SSEA-3 Rat anti-Human, Mouse, Biotin, Clone: eBioMC-631 (MC-631), eBioscience™

Rat Monoclonal Antibody

$183.48 - $368.28

Specifications

Antigen SSEA-3
Clone eBioMC-631 (MC-631)
Host Species Rat
Gene Alias stage-specific embryonic antigen-3
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012221 Life Technologies
LS13883380
25μg Each for $183.48
5012222 Life Technologies
LS13883382
100μg Each for $368.28
Specifications

Description

The eBioMC-631 (MC-631) monoclonal antibody reacts with the carbohydrate moiety galactosylgloboside on Stage-specific embryonic antigen-3 (SSEA-3). Expression is restricted to the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES) and down regulated following differentiation.
Description & Specifications

Specifications

SSEA-3
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eBioMC-631 (MC-631)
stage-specific embryonic antigen-3
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer