Please login to your online account to display your discounted pricing

SSEA-1 Mouse anti-Human, Mouse, PerCP-eFluor 710, Clone: eBioMC-480 (MC-480), eBioscience™

Mouse Monoclonal Antibody

$201.55 - $506.05

Specifications

Antigen SSEA-1
Clone eBioMC-480 (MC-480)
Host Species Murine
Gene Alias stage-specific embryonic antigen-1
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123915

 
life technologies
LS46881341
25 Tests Each for $201.55

501123916

 
life technologies
LS46881342
100 Tests Each for $506.05
Description & Specifications

Specifications

Antigen SSEA-1
Clone eBioMC-480 (MC-480)
Host Species Murine
Gene Alias stage-specific embryonic antigen-1
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The eBioMC-480 (MC-480) antibody reacts with the stage-specific embryonic antigen-1 (SSEA-1), a carbohydrate epitope expressed upon the surface of early mouse embryos, murine embryonal carcinoma cells (EC), murine embryonic stem cells (ES) and murine and human germ cells (EG). No immunoreactivity is evident with undifferentiated human EC and ES cells. Differentiation of human EC results in an increase in SSEA-1 expression, while in the mouse expression is diminished. Expression of the carbohydrate moiety is also found on mature human granulocytes (on CD15) and some monocytes. SSEA-1 is associated with cell adhesion, migration and differentiation.