Please login to your online account to display your discounted pricing

SSEA-1 Mouse anti-Human, Mouse, PE, Clone: eBioMC-480 (MC-480), eBioscience™

Mouse Monoclonal Antibody

$187.05 - $419.05

Specifications

Antigen SSEA-1
Clone eBioMC-480 (MC-480)
Host Species Mouse
Gene Alias stage-specific embryonic antigen-1
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011215

 
life technologies
LS12881341
25 tests Each for $187.05

5011216

 
life technologies
LS12881342
100 tests Each for $419.05
Description & Specifications

Specifications

Antigen SSEA-1
Clone eBioMC-480 (MC-480)
Host Species Mouse
Gene Alias stage-specific embryonic antigen-1
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The eBioMC-480 (MC-480) antibody reacts with the stage-specific embryonic antigen-1 (SSEA-1), a carbohydrate epitope expressed upon the surface of early mouse embryos, murine embryonal carcinoma cells (EC), murine embryonic stem cells (ES) and murine & human germ cells (EG). No immunoreactivity is evident with undifferentiated human EC and ES cells. Differentiation of human EC results in an increase in SSEA-1 expression, while in the mouse expression is diminished. Expression of the carbohydrate moiety is also found on mature human granulocytes (on CD15) and some monocytes. SSEA-1 is associated with cell adhesion, migration and differentiation.