Please login to your online account to display your discounted pricing

SSC Buffer, 20X Powder Pack, ULTROL™ Grade, Calbiochem™ EMD Millipore

Each pack provides an adequate amount of material to make 1L of 20X concentrate that can be diluted to a 1X solution containing 150mM NaCl, 15mM sodium citrate, pH 8.0

$81.64 - $81.64

Chemical Identifiers

Analyte or Component Names NaCl, Sodium Citrate
Chemical Name or Material SSC Buffer
Concentration or Composition (by Analyte or Components) 1X solution contains 150mM NaCl, 15mM sodium citrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5677802PACK

 
emd millipore corporation
5677802PACK
Each for $81.64
Description & Specifications

Specifications

Quantity 2 Pk.
Concentration or Composition (by Analyte or Components) 1X solution contains 150mM NaCl, 15mM sodium citrate
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Chemical Name or Material SSC Buffer
Synonym Saline-Sodium Citrate Buffer
Solubility Information Partially soluble in cold water
Analyte or Component Names NaCl, Sodium Citrate
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).