Invitrogen™ SRPK2 Recombinant Human Protein DFS Item
SRPK2, along with SRPK1, is most likely the cellular protein kinase mediating HBV core protein phosphorylation during viral infection.
Manufacturer:  Invitrogen™ LSPV4060
 View more versions of this product
Catalog No. PV4060
Description
Description
SRPK2 represents an important host cell target for therapeutic intervention in HBV infection.
Specifications
Specifications
NP_872633
Frozen
105kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
SRPK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
SRPK2, SFRSK2
Baculovirus
Human
GST Tag
Documents