Invitrogen™ SRMS (SRM) Recombinant Human Protein DFS Item
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family).
Manufacturer:  Invitrogen™ LSPV4287
 View more versions of this product
Catalog No. PV4287
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
SRMS (Srm) may be involved in the proliferation and differentiation of keratinocytes.
Specifications
Specifications
NP_543013
Frozen
82kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
SRC Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM HEPES pH 7.0, 150mM KCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
SRMS (Srm)
Baculovirus
Human
GST Tag
Documents