SRC Recombinant Human Protein DFS Item
SRC Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3584
 View more versions of this product
Catalog No. PV3584
Description
Description
SRC is a cytoplasmic tyrosine kinase and proto-oncogene involved in the regulation of embryonic development and cell growth.
Specifications
Specifications
NP_005408
Protein assays and analysis
Frozen
SRC Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
SRC
SRC was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 10% glycerol, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
SRC Recombinant Human Protein
Human
0.36mg/mL
Frozen
62.3kDa
90% by SDS-PAGE
–80°C
1 Each
SRC Recombinant Human Protein
Kinase assay
SRC
1940nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
His Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.