missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Src I1

Dual site Src kinase inhibitor

$218.74 - $915.10

Specifications

Product Type Src I1
Inhibitors Src I1
Molecular Weight 373.4
Purity >99
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
364210R Tocris™
364210
10mg Each for $218.74
364250R Tocris™
364250
50mg Each for $915.10
Specifications

Description

Potent, competitive dual site (ATP- and peptide-binding) Src kinase inhibitor (IC50 values are 44 and 88nM for Src and Lck respectively). Inhibits VEGFR2 and c-fms at higher concentrations (IC50 values are 0.32 and 30μM respectively). Can be used in parallel with PP 1 (Cat. No. 1397) and PP 2 (Cat. No. 1407) to inhibit Src family kinases.
  • Chemical Name: 6,7-Dimethoxy-N-(4-phenoxyphenyl)-4-quinazolinamine
  • Formula: C22H19N3O3
  • CAS Number: 179248-59-0
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

Src I1
Src I1
373.4
>99