Please login to your online account to display your discounted pricing

SR 95531 hydrobromide, Tocris

Selective, competitive GABAA receptor antagonist

$163.07 - $640.56

Chemical Identifiers

Chemical Name or Material SR 95531 hydrobromide
Assay Percent Range >98%
Molecular Formula C15H17N3O3·HBr
Formula Weight 368.23
Synonym 6-Imino-3-(4-methoxyphenyl)-1(6H)-pyridazinebutanoic acid hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

126250

 
r & d systems
126250
50mg Each for $640.56

126210

 
r & d systems
126210
10mg Each for $163.07
Description & Specifications

Specifications

Formula Weight 368.23
Quantity 50mg
Molecular Formula C15H17N3O3·HBr
Chemical Name or Material SR 95531 hydrobromide
CAS 104104-50-9
Synonym 6-Imino-3-(4-methoxyphenyl)-1(6H)-pyridazinebutanoic acid hydrobromide
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature