Please login to your online account to display your discounted pricing

SR 59230A hydrochloride, Tocris

Potent and selective β3 antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material SR 59230A hydrochloride
Assay Percent Range >98%
Molecular Formula C21H27NO2·HCl
Formula Weight 361.91
Synonym 1-(2-Ethylphenoxy)-3-[[(1S)-1,2,3,4-tetrahydro-1-naphthalenyl]amino]-(2S)-2-propanol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

151110

 
r & d systems
RDS151110
10mg Each for $239.63

151150

 
r & d systems
RDS151150
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 361.91
Quantity 10mg
Chemical Name or Material SR 59230A hydrochloride
Assay Percent Range >98%
CAS 1135278-41-9
Synonym 1-(2-Ethylphenoxy)-3-[[(1S)-1,2,3,4-tetrahydro-1-naphthalenyl]amino]-(2S)-2-propanol hydrochloride
Molecular Formula C21H27NO2·HCl
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature