Please login to your online account to display your discounted pricing

SR 58611A hydrochloride, Tocris™

β3 agonist; displays anxiolytic and antidepressant effects

$233.76 - $963.39

Chemical Identifiers

Chemical Name or Material SR 58611A hydrochloride
Assay Percent Range >98%
Molecular Formula C22H26ClNO4·HCl
Formula Weight 440.36
Synonym [[(7S)-7-[[(2R)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]-5,6,7,8-tetrahydro-2-naphthalenyl]ox]yacetic acid ethyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

398650

 
r & d systems
RDS398650
50mg Each for $963.39

398610

 
r & d systems
RDS398610
10mg Each for $233.76
Description & Specifications

Specifications

Chemical Name or Material SR 58611A hydrochloride
Quantity 50mg
CAS 121524-09-2
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >98%
Molecular Formula C22H26ClNO4·HCl
Formula Weight 440.36
Synonym [[(7S)-7-[[(2R)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]-5,6,7,8-tetrahydro-2-naphthalenyl]ox]yacetic acid ethyl ester hydrochloride
Safety and Handling

Desiccate at Room Temperature